4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine

Lieferant: Apollo Scientific

OR322194-250MG
APOSOR322194-250MGEA 58 EUR
APOSOR322194-250MG
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine
4-Chloro-5,6-dimethylthieno[2,3-d]pyrimidine
Formel: C₈H₇ClN₂S
Molecular Weight: 198.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00464832
CAS-Nummer: 108831-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR