6-Methoxy-m-kresol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methoxy-5-methylphenol, 2-Hydroxy-4-methylanisol, 3-Hydroxy-4-methoxytoluol, 1-Hydroxy-2-methoxy-5-methylbenzol
OR322195-25G
APOSOR322195-25GEA
325
EUR
APOSOR322195-25G
6-Methoxy-m-kresol 98%
6-Methoxy-m-kresol
Formel:
C₈H₁₀O₂ Molecular Weight: 138.17 g/mol Siedepunkt: 115…117 °C Schmelzpunkt: 35…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040901 CAS-Nummer: 1195-09-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...