(2E)-1-(Furan-3-yl)-3-[(4-methoxyphenyl)amino]prop-2-en-1-one, tech

Lieferant: Apollo Scientific

OR32221-1G
APOSOR32221-1GEA 1570 EUR
APOSOR32221-1G
(2E)-1-(Furan-3-yl)-3-[(4-methoxyphenyl)amino]prop-2-en-1-one, tech
(2E)-1-(Furan-3-yl)-3-[(4-methoxyphenyl)amino]prop-2-en-1-one
Formel: C₁₄H₁₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02082365
CAS-Nummer: 477854-96-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR