4-Chloro-2-methyl-5,6,7,8-tetrahydro[1]benzothieno[2,3-d]pyrimidine

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-methyl-5,6,7,8-tetrahydrobenzo[4,5]thieno[2,3-d]pyrimidine

OR322216-250MG
APOSOR322216-250MGEA 69.6 EUR
APOSOR322216-250MG
4-Chloro-2-methyl-5,6,7,8-tetrahydro[1]benzothieno[2,3-d]pyrimidine
4-Chloro-2-methyl-5,6,7,8-tetrahydro[1]benzothieno[2,3-d]pyrimidine
Formel: C₁₁H₁₁ClN₂S
Molecular Weight: 238,736 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00458475
CAS-Nummer: 81765-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR