Methyl 5-chloro-1,3-dimethyl-1H-pyrazole-4-carboxylate 95%
Lieferant: Apollo Scientific
OR32222-1G
APOSOR32222-1GEA
671
EUR
APOSOR32222-1G
Methyl 5-chloro-1,3-dimethyl-1H-pyrazole-4-carboxylate 95%
Methyl 5-chloro-1,3-dimethyl-1H-pyrazole-4-carboxylate
Formel:
C₇H₉ClN₂O₂ Molecular Weight: 188,612 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04124308 CAS-Nummer: 27006-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...