5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbonitrile 95%
Lieferant: Apollo Scientific
OR32223-1G
APOSOR32223-1GEA
671
EUR
APOSOR32223-1G
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbonitrile 95%
5-Chloro-1,3-dimethyl-1H-pyrazole-4-carbonitrile
Formel:
C₆H₆ClN₃ Molecular Weight: 155,585 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569082 CAS-Nummer: 96286-02-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...