1,3-Bis(methoxycarbonyl)-2-methyl-2-thiopseudourea
Lieferant: Apollo Scientific
|
Achtung
|
OR322230-5G
APOSOR322230-5GEA
0
EUR
APOSOR322230-5G
1,3-Bis(methoxycarbonyl)-2-methyl-2-thiopseudourea
1,3-Bis(methoxycarbonyl)-2-methyl-2-thiopseudourea
Formel:
C₆H₁₀N₂O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00674104 CAS-Nummer: 34840-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...