cis-4-Cyclopentene-1,3-diol

Lieferant: Apollo Scientific

Synonyme: cis-Cyclopent-4-ene-1,3-diol

OR322236-100MG
APOSOR322236-100MGEA 79.9 EUR
APOSOR322236-100MG
cis-4-Cyclopentene-1,3-diol
cis-4-Cyclopentene-1,3-diol
Formel: C₅H₈O₂
Molecular Weight: 100,116 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077703
CAS-Nummer: 29783-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR