3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-(prop-2-en-1-yl)-1,2-dihydropyridin-2-one, tech

Lieferant: Apollo Scientific

OR32224-1G
APOSOR32224-1GEA 1570 EUR
APOSOR32224-1G
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-(prop-2-en-1-yl)-1,2-dihydropyridin-2-one, tech
3-[(4-Chlorophenyl)methyl]-4-hydroxy-1-(prop-2-en-1-yl)-1,2-dihydropyridin-2-one
Formel: C₁₅H₁₄ClNO₂
Molecular Weight: 275,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01871428
CAS-Nummer: 477856-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR