5-(Methylthio)-1,2,4-thiadiazol-3-amine

Lieferant: Apollo Scientific

Synonyme: 3-Amino-5-methylthio-1,2,4-thiadiazole

OR322242-1G
APOSOR322242-1GEA 0 EUR
APOSOR322242-1G
5-(Methylthio)-1,2,4-thiadiazol-3-amine
5-(Methylthio)-1,2,4-thiadiazol-3-amine
Formel: C₃H₅N₃S₂
Molecular Weight: 147,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00799288
CAS-Nummer: 60093-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR