4-Chloro-2-[(E)-[(dimethylamino)methylidene]amino]benzoic acid, tech
Lieferant: Apollo Scientific
OR32225-1G
APOSOR32225-1GEA
0
EUR
APOSOR32225-1G
4-Chloro-2-[(E)-[(dimethylamino)methylidene]amino]benzoic acid, tech
4-Chloro-2-[(E)-[(dimethylamino)methylidene]amino]benzoic acid
Formel:
C₁₀H₁₁ClN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01871429 CAS-Nummer: 477856-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...