Methyl 1-(4-chlorobenzoyl)piperidine-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32226-1G
APOSOR32226-1GEA 1570 EUR
APOSOR32226-1G
Methyl 1-(4-chlorobenzoyl)piperidine-4-carboxylate, tech
Methyl 1-(4-chlorobenzoyl)piperidine-4-carboxylate
Formel: C₁₄H₁₆ClNO₃
Molecular Weight: 281,735 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082485
CAS-Nummer: 349614-05-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR