Methyl 1-(4-methoxybenzoyl)piperidine-4-carboxylate, tech

Lieferant: Apollo Scientific

OR32227-1G
APOSOR32227-1GEA 1570 EUR
APOSOR32227-1G
Methyl 1-(4-methoxybenzoyl)piperidine-4-carboxylate, tech
Methyl 1-(4-methoxybenzoyl)piperidine-4-carboxylate
Formel: C₁₅H₁₉NO₄
Molecular Weight: 277,316 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01568754
CAS-Nummer: 303994-55-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR