7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
7-Chloro-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one, 7-Chloro-1,2,3,4-tetrahydro-5H-1-benzazepin-5-one
OR322271-1G
APOSOR322271-1GEA
21.9
EUR
APOSOR322271-1G
7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one 98%
7-Chloro-1,2,3,4-tetrahydrobenzo[b]azepin-5-one
Formel:
C₁₀H₁₀ClNO Molecular Weight: 195,6455 g/mol Schmelzpunkt: 105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06738694 CAS-Nummer: 160129-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...