3-Bromo-4-chlorobenzyl alcohol 97%

Lieferant: Apollo Scientific
Achtung

OR322285-1G
APOSOR322285-1GEA 21.9 EUR
APOSOR322285-1G
3-Bromo-4-chlorobenzyl alcohol 97%
3-Bromo-4-chlorobenzyl alcohol
Formel: C₇H₆BrClO
Molecular Weight: 221.479 g/mol
Siedepunkt: 100 °C (0.15 mm Hg)
Lagertemperatur: Raumtemperatur
MDL: MFCD08059085
CAS-Nummer: 329218-12-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR