Dansylamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Dimethylamino)naphthalinsulfonamid, 5-(Dimethylamino)naphthalin-1-sulfonamid, 5-(Dimethylamino)naphthalinsulfonsäureamid
OR322291-1G
APOSOR322291-1GEA
149
EUR
APOSOR322291-1G
Dansylamid
Dansylamid
Formel:
C₁₂H₁₄N₂O₂S Molecular Weight: 250,32 g/mol Schmelzpunkt: 219 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004000 CAS-Nummer: 1431-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...