Boc-4-phenylpiperidine-4-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR322299-250MG
APOSOR322299-250MGEA 26.1 EUR
APOSOR322299-250MG
Boc-4-phenylpiperidine-4-carboxylic acid
Boc-4-phenylpiperidine-4-carboxylic acid
Formel: C₁₇H₂₃NO₄
Molecular Weight: 305.38 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00673774
CAS-Nummer: 167262-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR