DL-2-Methylaminosuccinic acid

Lieferant: Apollo Scientific
Achtung

OR322308-1G
APOSOR322308-1GEA 0 EUR
APOSOR322308-1G
DL-2-Methylaminosuccinic acid
DL-2-Methylaminosuccinic acid
Formel: C₅H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00065918
CAS-Nummer: 17833-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR