4-tert-Butyl-2-[(1Z)-(hydroxyimino)methyl]phenol, tech
Lieferant: Apollo Scientific
OR32231-1G
APOSOR32231-1GEA
1570
EUR
APOSOR32231-1G
4-tert-Butyl-2-[(1Z)-(hydroxyimino)methyl]phenol, tech
4-tert-Butyl-2-[(1Z)-(hydroxyimino)methyl]phenol
Formel:
C₁₁H₁₅NO₂ Molecular Weight: 193,242 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01568847 CAS-Nummer: 50849-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...