2-Chloro-2-(phenylhydrazono)acetic acid ethyl ester
Lieferant: Apollo Scientific
Synonyme: Ethyl 2-chloro-2-(2-phenylhydrazono)acetate
OR322310-1G
APOSOR322310-1GEA 82.8 EUR
APOSOR322310-1G
2-Chloro-2-(phenylhydrazono)acetic acid ethyl ester
2-Chloro-2-(phenylhydrazono)acetic acid ethyl ester
Formel: C₁₀H₁₁ClN₂O₂
Molecular Weight: 226,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00228707
CAS-Nummer: 28663-68-5