4-Chloro-6-(1h-imidazol-1-yl)pyrimidine

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-6-(1H-imidazol-1-yl)pyrimidine

OR322318-250MG
APOSOR322318-250MGEA 53.8 EUR
APOSOR322318-250MG
4-Chloro-6-(1h-imidazol-1-yl)pyrimidine
4-Chloro-6-(1h-imidazol-1-yl)pyrimidine
Formel: C₇H₅ClN₄
Molecular Weight: 180,594 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01631197
CAS-Nummer: 114834-02-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR