3',4'-Dimethylacetophenon

Lieferant: Apollo Scientific

Synonyme: 1-(3,4-Dimethylphenyl)ethan-1-on

OR322319-25G
APOSOR322319-25GEA 101 EUR
APOSOR322319-25G
3',4'-Dimethylacetophenon
3',4'-Dimethylacetophenon
Formel: C₁₀H₁₂O
Molecular Weight: 148,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008743
CAS-Nummer: 3637-01-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR