3-Bromo-4-(morpholin-4-yl)aniline 95%

Lieferant: Apollo Scientific

OR32233-1G
APOSOR32233-1GEA 156 EUR
APOSOR32233-1G
3-Bromo-4-(morpholin-4-yl)aniline 95%
3-Bromo-4-(morpholin-4-yl)aniline
Formel: C₁₀H₁₃BrN₂O
Molecular Weight: 257.127 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02081966
CAS-Nummer: 347324-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR