4-[4-(Pyridin-2-yl)pyrimidin-2-yl]phenol, tech

Lieferant: Apollo Scientific

OR32234-1G
APOSOR32234-1GEA 1570 EUR
APOSOR32234-1G
4-[4-(Pyridin-2-yl)pyrimidin-2-yl]phenol, tech
4-[4-(Pyridin-2-yl)pyrimidin-2-yl]phenol
Formel: C₁₅H₁₁N₃O
Molecular Weight: 249,267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187283
CAS-Nummer: 477856-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR