5-Chlor-2-nitro-p-toluidin 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-4-methyl-2-nitroaniline

OR322344-25G
APOSOR322344-25GEA 195 EUR
APOSOR322344-25G
5-Chlor-2-nitro-p-toluidin 95%
5-Chlor-2-nitro-p-toluidin
Formel: C₇H₇ClN₂O₂
Molecular Weight: 186.60 g/mol
Schmelzpunkt: 159…163 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00034066
CAS-Nummer: 7149-80-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR