1,2-Dimethyl (1R,2R)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech

Lieferant: Apollo Scientific

OR32236-1G
APOSOR32236-1GEA 1570 EUR
APOSOR32236-1G
1,2-Dimethyl (1R,2R)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate, tech
1,2-Dimethyl (1R,2R)-4,5-dimethylcyclohex-4-ene-1,2-dicarboxylate
Formel: C₁₂H₁₈O₄
Molecular Weight: 226,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08690904
CAS-Nummer: 1881275-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR