6-Brom-3-methoxysalicylaldehyd
Lieferant: Apollo Scientific
Synonyme:
6-Brom-2-hydroxy-3-methoxybenzaldehyd
OR322369-1G
APOSOR322369-1GEA
27.7
EUR
APOSOR322369-1G
6-Brom-3-methoxysalicylaldehyd
6-Brom-3-methoxysalicylaldehyd
Formel:
C₈H₇BrO₃ Molecular Weight: 231,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046152 CAS-Nummer: 20035-41-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...