2-Bromethyl-phenylsulfid
Lieferant: Apollo Scientific
Synonyme:
(2-Bromoethyl)(phenyl)sulphane
OR322375-5G
APOSOR322375-5GEA
68.3
EUR
APOSOR322375-5G
2-Bromethyl-phenylsulfid
2-Bromethyl-phenylsulfid
Formel:
C₈H₉BrS Molecular Weight: 217,12 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040826 CAS-Nummer: 4837-01-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...