4-((Dimethylamino)methyl)aniline
Lieferant: Apollo Scientific
Synonyme:
4-Amino-N,N-dimethylbenzylamine
OR322378-5G
APOSOR322378-5GEA
95.7
EUR
APOSOR322378-5G
4-((Dimethylamino)methyl)aniline
4-((Dimethylamino)methyl)aniline
Formel:
C₉H₁₄N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035953 CAS-Nummer: 6406-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...