5-Methoxy-1-benzofuran-2-carbonitrile, tech

Lieferant: Apollo Scientific

OR32238-1G
APOSOR32238-1GEA 1570 EUR
APOSOR32238-1G
5-Methoxy-1-benzofuran-2-carbonitrile, tech
5-Methoxy-1-benzofuran-2-carbonitrile
Formel: C₁₀H₇NO₂
Molecular Weight: 173.168 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02187499
CAS-Nummer: 35351-47-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR