4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid 95%
Lieferant: Apollo Scientific
OR32241-1G
OR32241-5G
APOSOR32241-1GEA
180
EUR
APOSOR32241-1G
APOSOR32241-5G
4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid 95%
4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid
Formel:
C₁₄H₁₇NO₃ Molecular Weight: 247.3 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02082683 CAS-Nummer: 298700-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...