2-(1-Hydroxy-2-methylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one, tech

Lieferant: Apollo Scientific

OR32242-1G
APOSOR32242-1GEA 1570 EUR
APOSOR32242-1G
2-(1-Hydroxy-2-methylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one, tech
2-(1-Hydroxy-2-methylpropan-2-yl)-2,3-dihydro-1H-isoindol-1-one
Formel: C₁₂H₁₅NO₂
Molecular Weight: 205,253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02082781
CAS-Nummer: 107400-33-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR