1-Benzyl-4-(ethoxycarbonylmethyl)piperazine

Lieferant: Apollo Scientific
Achtung

OR322421-500MG
APOSOR322421-500MGEA 0 EUR
APOSOR322421-500MG
1-Benzyl-4-(ethoxycarbonylmethyl)piperazine
1-Benzyl-4-(ethoxycarbonylmethyl)piperazine
Formel: C₁₅H₂₂N₂O₂
Siedepunkt: 155…160 °C/0,8mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01076590
CAS-Nummer: 23173-76-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR