Cyclobutylamine hydrochloride

Lieferant: Apollo Scientific

OR322423-5G
APOSOR322423-5GEA 0 EUR
APOSOR322423-5G
Cyclobutylamine hydrochloride
Cyclobutylamine hydrochloride
Formel: C₄H₉N·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00034953
CAS-Nummer: 6291-01-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR