3-Acetylaminobenzenesulphonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR322436-1G
OR322436-250MG
APOSOR322436-1GEA
0
EUR
APOSOR322436-1G
APOSOR322436-250MG
3-Acetylaminobenzenesulphonyl chloride
3-Acetylaminobenzenesulphonyl chloride
Formel:
C₈H₈ClNO₃S Schmelzpunkt: 88…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02222873 CAS-Nummer: 23905-46-6 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...