2,5-Dimethyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine 95%

Lieferant: Apollo Scientific

OR32244-1G OR32244-5G
APOSOR32244-1GEA 180 EUR
APOSOR32244-1G APOSOR32244-5G
2,5-Dimethyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine 95%
2,5-Dimethyl-6-phenylpyrazolo[1,5-a]pyrimidin-7-amine
Formel: C₁₄H₁₄N₄
Molecular Weight: 238.288 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02661827
CAS-Nummer: 428845-73-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR