2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-amine 95%

Lieferant: Apollo Scientific

OR32245-1G
APOSOR32245-1GEA 427 EUR
APOSOR32245-1G
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-amine 95%
2,5-Dimethylpyrazolo[1,5-a]pyrimidin-7-amine
Formel: C₈H₁₀N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00995337
CAS-Nummer: 2369-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR