5-(1-Pyrrolidinyl)-2-furaldehyde
Lieferant: Apollo Scientific
OR322455-250MG
APOSOR322455-250MGEA
74.1
EUR
APOSOR322455-250MG
5-(1-Pyrrolidinyl)-2-furaldehyde
5-(1-Pyrrolidinyl)-2-furaldehyde
Formel:
C₉H₁₁NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02629619 CAS-Nummer: 84966-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...