4-Methyl-1H,2H,3H,5H,9bH-indeno[1,2-d]pyrimidine-2,5-dione, tech

Lieferant: Apollo Scientific

OR32247-1G
APOSOR32247-1GEA 1570 EUR
APOSOR32247-1G
4-Methyl-1H,2H,3H,5H,9bH-indeno[1,2-d]pyrimidine-2,5-dione, tech
4-Methyl-1H,2H,3H,5H,9bH-indeno[1,2-d]pyrimidine-2,5-dione
Formel: C₁₂H₁₀N₂O₂
Molecular Weight: 214,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02572010
CAS-Nummer: 123044-21-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR