4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine
Lieferant: Apollo Scientific
OR322475-500MG
APOSOR322475-500MGEA
109
EUR
APOSOR322475-500MG
4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine
4-Chloro-6,7-dihydro-5H-cyclopenta[4,5]thieno[2,3-d]pyrimidine
Formel:
C₉H₇ClN₂S Molecular Weight: 210,69 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00572400 CAS-Nummer: 300816-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...