2-(1-Methylhydrazino)-4,5-dihydro-1H-imidazole hydrobromide
Lieferant: Apollo Scientific
Synonyme:
N-(4,5-Dihydroimidazol-2-yl)-n-methylhydrazine hydrobromide
OR322478-1G
APOSOR322478-1GEA
0
EUR
APOSOR322478-1G
2-(1-Methylhydrazino)-4,5-dihydro-1H-imidazole hydrobromide
2-(1-Methylhydrazino)-4,5-dihydro-1H-imidazole hydrobromide
Formel:
C₄H₁₁BrN₄ Molecular Weight: 195,07 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00067859 CAS-Nummer: 55959-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...