6-Chloro-3-nitropyridine-2-carboxamide

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-3-nitropicolinamide

OR322491-250MG
APOSOR322491-250MGEA 85.7 EUR
APOSOR322491-250MG
6-Chloro-3-nitropyridine-2-carboxamide
6-Chloro-3-nitropyridine-2-carboxamide
Formel: C₆H₄ClN₃O₃
Molecular Weight: 201,567 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07780776
CAS-Nummer: 171178-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR