2-Aminopyrimidine-5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Aminopyrimidine-5-carboxaldehyde

OR322504-5G
APOSOR322504-5GEA 94.4 EUR
APOSOR322504-5G
2-Aminopyrimidine-5-carbaldehyde
2-Aminopyrimidine-5-carbaldehyde
Formel: C₅H₅N₃O
Molecular Weight: 123.11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701714
CAS-Nummer: 120747-84-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR