2-Methylveratraldehyde

Lieferant: Apollo Scientific

Synonyme: 3,4-Dimethoxy-2-methylbenzaldehyde

OR322506-250MG
APOSOR322506-250MGEA 62.5 EUR
APOSOR322506-250MG
2-Methylveratraldehyde
2-Methylveratraldehyde
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03428551
CAS-Nummer: 51234-09-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR