2-Phenyl-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific

OR322513-5G
APOSOR322513-5GEA 0 EUR
APOSOR322513-5G
2-Phenyl-1H-indole-3-carbaldehyde
2-Phenyl-1H-indole-3-carbaldehyde
Formel: C₁₅H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00435481
CAS-Nummer: 25365-71-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR