Methyl-4-acetyl-1H-pyrrole-2-carboxylate 95%

Lieferant: Apollo Scientific

OR32252-1G
APOSOR32252-1GEA 488 EUR
APOSOR32252-1G
Methyl-4-acetyl-1H-pyrrole-2-carboxylate 95%
Methyl-4-acetyl-1H-pyrrole-2-carboxylate
Formel: C₈H₉NO₃
Molecular Weight: 167,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02571735
CAS-Nummer: 40611-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR