Ethyl-2-nitrobenzoylacetate

Lieferant: Apollo Scientific

Synonyme: Benzenepropanoic acid,2-nitro-b-oxo-, ethyl ester, ethyl 3-(2-nitrophenyl)-3-oxopropanoate, 2-Nitrobenzoylacetic acid ethyl ester

OR322527-1G
APOSOR322527-1GEA 126 EUR
APOSOR322527-1G
Ethyl-2-nitrobenzoylacetate
Ethyl-2-nitrobenzoylacetate
Formel: C₁₁H₁₁NO₅
Molecular Weight: 237,211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03424821
CAS-Nummer: 52119-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR