4-Chloro-6-methyl-2-(2-pyridinyl)pyrimidine 94%

Lieferant: Apollo Scientific

OR32253-1G
APOSOR32253-1GEA 393 EUR
APOSOR32253-1G
4-Chloro-6-methyl-2-(2-pyridinyl)pyrimidine 94%
4-Chloro-6-methyl-2-(2-pyridinyl)pyrimidine
Formel: C₁₀H₈ClN₃
Molecular Weight: 205,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00662786
CAS-Nummer: 77168-31-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR