(3E)-3-{1-[(2-Methoxyphenyl)amino]ethylidene}oxolan-2-one, tech

Lieferant: Apollo Scientific

OR32255-1G
APOSOR32255-1GEA 1570 EUR
APOSOR32255-1G
(3E)-3-{1-[(2-Methoxyphenyl)amino]ethylidene}oxolan-2-one, tech
(3E)-3-{1-[(2-Methoxyphenyl)amino]ethylidene}oxolan-2-one
Formel: C₁₃H₁₅NO₃
Molecular Weight: 233,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03617587
CAS-Nummer: 123438-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR