Ethyl 3-amino-1H-indole-2-carboxylate

Lieferant: Apollo Scientific

OR322552-250MG
APOSOR322552-250MGEA 133 EUR
APOSOR322552-250MG
Ethyl 3-amino-1H-indole-2-carboxylate
Ethyl 3-amino-1H-indole-2-carboxylate
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,225 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00141060
CAS-Nummer: 87223-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR